Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 105236973-105237122 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TET2
Entrez Gene ID
54790
Gene Name
tet methylcytosine dioxygenase 2
Gene Aliases
KIAA1546, MDS
UniGene
Hs.367639
Gene Chromosome Location
Chr.4: 105145875-105279803 on Build GRCh38
Transcripts
NM_001127208.2 NM_017628.4 XM_005263082.2 XM_006714242.3 XM_017008319.1 NR_126420.1

Gene Symbol
TET2-AS1
Entrez Gene ID
104384744
Gene Name
TET2 antisense RNA 1
Gene Aliases
-
UniGene
Hs.624109
Gene Chromosome Location
Chr.4: 105171354-105352881 on Build GRCh38
Transcripts
NM_001127208.2 NM_017628.4 XM_005263082.2 XM_006714242.3 XM_017008319.1 NR_126420.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4170110 c.3109_3110insC p.K1038fs*5 Chr.4: 105237051-105237052 on Build GRCh38
6912798 c.3040G>A p.A1014T Chr.4: 105236982-105236982 on Build GRCh38
4383892 c.3169G>T p.E1057* Chr.4: 105237111-105237111 on Build GRCh38
7283031 c.3160G>A p.V1054I Chr.4: 105237102-105237102 on Build GRCh38
3735042 c.3064_3065delAA p.K1022fs*4 Chr.4: 105237006-105237007 on Build GRCh38
View More

View Full Product Details