Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 105234671-105234800 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TET2
Entrez Gene ID
54790
Gene Name
tet methylcytosine dioxygenase 2
Gene Aliases
KIAA1546, MDS
UniGene
Hs.367639
Gene Chromosome Location
Chr.4: 105145875-105279803 on Build GRCh38
Transcripts
NM_001127208.2 NM_017628.4 XM_005263082.2 XM_006714242.3 XM_017008319.1 NR_126420.1

Gene Symbol
TET2-AS1
Entrez Gene ID
104384744
Gene Name
TET2 antisense RNA 1
Gene Aliases
-
UniGene
Hs.624109
Gene Chromosome Location
Chr.4: 105171354-105352881 on Build GRCh38
Transcripts
NM_001127208.2 NM_017628.4 XM_005263082.2 XM_006714242.3 XM_017008319.1 NR_126420.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5944490 c.836_837insAGAC p.S280fs*3 Chr.4: 105234778-105234779 on Build GRCh38
5762949 c.769A>G p.T257A Chr.4: 105234711-105234711 on Build GRCh38
5878929 c.729_730insA p.T245fs*9 Chr.4: 105234671-105234672 on Build GRCh38
144508 c.803C>G p.S268W Chr.4: 105234745-105234745 on Build GRCh38
4956767 c.749A>G p.N250S Chr.4: 105234691-105234691 on Build GRCh38
View More

View Full Product Details