Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 68228685-68228936 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMPRSS11B
Entrez Gene ID
132724
Gene Name
transmembrane protease, serine 11B
Gene Aliases
-
UniGene
Hs.407122
Gene Chromosome Location
Chr.4: 68226653-68245694 on Build GRCh38
Transcripts
NM_182502.3 XM_011531608.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5908453 c.1089G>A p.Q363Q Chr.4: 68228742-68228742 on Build GRCh38
1056818 c.1074A>G p.E358E Chr.4: 68228757-68228757 on Build GRCh38
4125630 c.1048T>C p.L350L Chr.4: 68228783-68228783 on Build GRCh38
6804523 c.998A>G p.K333R Chr.4: 68228833-68228833 on Build GRCh38
258208 c.970G>T p.E324* Chr.4: 68228861-68228861 on Build GRCh38
View More

View Full Product Details