Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 68232253-68232526 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMPRSS11B
Entrez Gene ID
132724
Gene Name
transmembrane protease, serine 11B
Gene Aliases
-
UniGene
Hs.407122
Gene Chromosome Location
Chr.4: 68226653-68245694 on Build GRCh38
Transcripts
NM_182502.3 XM_011531608.2 XM_011531609.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1430632 c.499A>G p.T167A Chr.4: 68232387-68232387 on Build GRCh38
345651 c.504C>A p.N168K Chr.4: 68232382-68232382 on Build GRCh38
1619006 c.470-5T>A p.? Chr.4: 68232421-68232421 on Build GRCh38

View Full Product Details