Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 68234358-68234615 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMPRSS11B
Entrez Gene ID
132724
Gene Name
transmembrane protease, serine 11B
Gene Aliases
-
UniGene
Hs.407122
Gene Chromosome Location
Chr.4: 68226653-68245694 on Build GRCh38
Transcripts
NM_182502.3 XM_011531608.2 XM_011531609.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5569182 c.385A>G p.T129A Chr.4: 68234547-68234547 on Build GRCh38
354421 c.336G>C p.Q112H Chr.4: 68234596-68234596 on Build GRCh38
1291946 c.318C>T p.A106A Chr.4: 68234614-68234614 on Build GRCh38
4734693 c.365C>T p.A122V Chr.4: 68234567-68234567 on Build GRCh38
6167692 c.443C>A p.A148E Chr.4: 68234489-68234489 on Build GRCh38
View More

View Full Product Details