Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 19376120-19376393 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RPS6
Entrez Gene ID
6194
Gene Name
ribosomal protein S6
Gene Aliases
S6
UniGene
Hs.408073
Gene Chromosome Location
Chr.9: 19376256-19380237 on Build GRCh38
Transcripts
NM_001010.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1107651 c.698G>T p.R233I Chr.9: 19376345-19376345 on Build GRCh38
422403 c.690G>C p.K230N Chr.9: 19376353-19376353 on Build GRCh38
1314707 c.666G>C p.E222D Chr.9: 19376377-19376377 on Build GRCh38
1107652 c.695G>A p.R232H Chr.9: 19376348-19376348 on Build GRCh38
4373343 c.686C>T p.A229V Chr.9: 19376357-19376357 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372915420 Chr.9: 19376338-19376338 on Build GRCh38
rs1049779861 Chr.9: 19376169-19376169 on Build GRCh38
rs780806374 Chr.9: 19376331-19376331 on Build GRCh38
rs756828696 Chr.9: 19376322-19376322 on Build GRCh38
rs778266210 Chr.9: 19376356-19376356 on Build GRCh38
rs553421652 Chr.9: 19376290-19376290 on Build GRCh38
rs768929394 Chr.9: 19376277-19376277 on Build GRCh38
rs369681273 Chr.9: 19376264-19376264 on Build GRCh38
rs765126928 Chr.9: 19376265-19376269 on Build GRCh38
rs745442351 Chr.9: 19376271-19376271 on Build GRCh38
rs1055228895 Chr.9: 19376259-19376259 on Build GRCh38
rs748611445 Chr.9: 19376348-19376348 on Build GRCh38
rs994439195 Chr.9: 19376242-19376242 on Build GRCh38
rs779539344 Chr.9: 19376339-19376339 on Build GRCh38
rs897066932 Chr.9: 19376218-19376218 on Build GRCh38
rs67906804 Chr.9: 19376260-19376260 on Build GRCh38
rs772310064 Chr.9: 19376352-19376352 on Build GRCh38
rs750119568 Chr.9: 19376334-19376334 on Build GRCh38
rs758411939 Chr.9: 19376247-19376247 on Build GRCh38
rs754802484 Chr.9: 19376237-19376237 on Build GRCh38
rs937704805 Chr.9: 19376216-19376216 on Build GRCh38
rs113373506 Chr.9: 19376158-19376158 on Build GRCh38
rs562166683 Chr.9: 19376178-19376178 on Build GRCh38
rs10119465 Chr.9: 19376146-19376146 on Build GRCh38
rs758126822 Chr.9: 19376314-19376314 on Build GRCh38
rs753550645 Chr.9: 19376245-19376254 on Build GRCh38
rs750510123 Chr.9: 19376316-19376321 on Build GRCh38
rs777831669 Chr.9: 19376290-19376294 on Build GRCh38
rs746587364 Chr.9: 19376256-19376256 on Build GRCh38
rs776871563 Chr.9: 19376303-19376303 on Build GRCh38
rs1023360483 Chr.9: 19376304-19376304 on Build GRCh38
rs927621630 Chr.9: 19376206-19376206 on Build GRCh38
rs933881985 Chr.9: 19376266-19376266 on Build GRCh38
rs757445661 Chr.9: 19376268-19376271 on Build GRCh38
rs773586781 Chr.9: 19376281-19376281 on Build GRCh38
rs772673878 Chr.9: 19376279-19376279 on Build GRCh38
rs781008957 Chr.9: 19376258-19376258 on Build GRCh38
rs3209385 Chr.9: 19376371-19376371 on Build GRCh38
rs759052038 Chr.9: 19376261-19376265 on Build GRCh38
rs941353754 Chr.9: 19376175-19376175 on Build GRCh38
rs759707353 Chr.9: 19376308-19376308 on Build GRCh38
rs889798806 Chr.9: 19376171-19376171 on Build GRCh38
rs758470032 Chr.9: 19376267-19376268 on Build GRCh38
rs754607550 Chr.9: 19376368-19376368 on Build GRCh38
rs562946854 Chr.9: 19376278-19376278 on Build GRCh38
rs1038272670 Chr.9: 19376212-19376212 on Build GRCh38
rs560053449 Chr.9: 19376357-19376357 on Build GRCh38
rs760802711 Chr.9: 19376282-19376282 on Build GRCh38
rs1143622 Chr.9: 19376318-19376318 on Build GRCh38
rs67710536 Chr.9: 19376257-19376257 on Build GRCh38
rs765357060 Chr.9: 19376310-19376310 on Build GRCh38
rs115304926 Chr.9: 19376167-19376167 on Build GRCh38
rs3206714 Chr.9: 19376367-19376367 on Build GRCh38
rs11541612 Chr.9: 19376330-19376330 on Build GRCh38
rs367650988 Chr.9: 19376241-19376241 on Build GRCh38
rs1012019872 Chr.9: 19376292-19376292 on Build GRCh38
rs756253094 Chr.9: 19376344-19376365 on Build GRCh38
rs3209386 Chr.9: 19376344-19376344 on Build GRCh38
rs757388872 Chr.9: 19376299-19376299 on Build GRCh38

View Full Product Details