Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 105233875-105234114 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TET2
Entrez Gene ID
54790
Gene Name
tet methylcytosine dioxygenase 2
Gene Aliases
KIAA1546, MDS
UniGene
Hs.367639
Gene Chromosome Location
Chr.4: 105145875-105279803 on Build GRCh38
Transcripts
NM_001127208.2 NM_017628.4 XM_005263082.2 XM_006714242.3 XM_017008319.1 NR_126420.1

Gene Symbol
TET2-AS1
Entrez Gene ID
104384744
Gene Name
TET2 antisense RNA 1
Gene Aliases
-
UniGene
Hs.624109
Gene Chromosome Location
Chr.4: 105171354-105352881 on Build GRCh38
Transcripts
NM_001127208.2 NM_017628.4 XM_005263082.2 XM_006714242.3 XM_017008319.1 NR_126420.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3598848 c.137C>T p.P46L Chr.4: 105234079-105234079 on Build GRCh38
3598846 c.44G>T p.S15I Chr.4: 105233986-105233986 on Build GRCh38
6945809 c.133C>T p.H45Y Chr.4: 105234075-105234075 on Build GRCh38
3775458 c.105G>A p.Q35Q Chr.4: 105234047-105234047 on Build GRCh38
231546 c.65C>T p.P22L Chr.4: 105234007-105234007 on Build GRCh38
View More

View Full Product Details