Gene Transcripts
- Gene Symbol
- Bnip2
- Entrez Gene ID
- 12175
- Gene Name
- BCL2/adenovirus E1B interacting protein 2
- Gene Aliases
- 5730523P12Rik, BNIP2beta, Nip21
- Chromosome Location
- Chr. 9: 69989425 - 70011659 on Build GRCm38
- UniGene ID
- Mm.159777
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001008238.3 |
NP_001008239.1
|
3, 4 |
401 |
|
NM_016787.4 |
NP_058067.2
|
3, 4 |
401 |
|
GenBank mRNA |
AF035207.1 |
- |
3, 4 |
253 |
|
AF400107.1 |
- |
2, 3 |
141 |
|
AK014659.1 |
- |
3, 4 |
353 |
|
AK054288.1 |
- |
3, 4 |
367 |
|
AK159745.1 |
- |
3, 4 |
364 |
|
AK161116.1 |
- |
3, 4 |
354 |
|
AK169609.1 |
- |
3, 4 |
386 |
|
AY591757.1 |
- |
2, 3 |
120 |
|
AY591758.1 |
- |
2, 3 |
120 |
|
BC012670.1 |
- |
3, 4 |
360 |
4037627 |
View Full Product Details