Gene Transcripts

Gene Symbol
Wdr81
Entrez Gene ID
192652
Gene Name
WD repeat domain 81
Gene Aliases
BC054822, Gm883, mFLJ00182
Chromosome Location
Chr. 11: 75440943 - 75454717 on Build GRCm38
UniGene ID
Mm.340378
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138950.2 NP_620400.2 1 2012
GenBank mRNA AK217852.1 - 1 56

View Full Product Details