Gene Transcripts

Gene Symbol
Cntnap5c
Entrez Gene ID
620292
Gene Name
contactin associated protein-like 5C
Gene Aliases
Caspr5-3, EG620292, Gm325
Chromosome Location
Chr. 17: 57769077 - 58410429 on Build GRCm38
UniGene ID
Mm.391030
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081653.1 NP_001075122.1 4 506

View Full Product Details