Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_029629.2 | NP_083905.1 | 6 | 909 | |
GenBank mRNA | AI841210.1 | - | 3 | 330 | |
AK002291.1 | - | 6 | 887 | ||
AK005134.1 | - | 5 | 1146 | ||
AK080868.1 | - | 6 | 895 | ||
AK170875.1 | - | 6 | 883 | ||
BC030182.1 | - | 6 | 860 | 5044017 |