Gene Transcripts

Gene Symbol
1700016H13Rik
Entrez Gene ID
74218
Gene Name
RIKEN cDNA 1700016H13 gene
Gene Aliases
Chromosome Location
Chr. 5: 103648583 - 103655863 on Build GRCm38
UniGene ID
Mm.159141
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163550.1 NP_001157022.1 4 447
NM_028824.1 NP_083100.1 4 598
GenBank mRNA AK006033.1 - 4 600
BC049644.1 - 4 590 6705673

View Full Product Details