Gene Transcripts

Gene Symbol
Trip10
Entrez Gene ID
116717
Gene Name
thyroid hormone receptor interactor 10
Gene Aliases
Cip4
Chromosome Location
Chr. 9: 9689214 - 9702353 on Build Rnor_6.0
UniGene ID
Rn.10814
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053920.1 NP_446372.1 12 1473
GenBank mRNA AB006914.1 - 12 1473
BC061840.1 - 12 1475 5600051

View Full Product Details