Gene Transcripts

Gene Symbol
St13
Entrez Gene ID
81800
Gene Name
suppression of tumorigenicity 13
Gene Aliases
Chromosome Location
Chr. 7: 122585770 - 122635731 on Build Rnor_6.0
UniGene ID
Rn.119613
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031122.1 NP_112384.1 3 257
GenBank mRNA BC078804.1 - 3 264
FQ212434.1 - 314
FQ228746.1 - 3 219
X82021.1 - 3 257

View Full Product Details