Gene Transcripts

Gene Symbol
Capn10
Entrez Gene ID
63834
Gene Name
calpain 10
Gene Aliases
Chromosome Location
Chr. 9: 100104000 - 100112833 on Build Rnor_6.0
UniGene ID
Rn.176334
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031673.2 NP_113861.1 1 284
GenBank mRNA AB113362.1 - 1 191
AB113363.1 - 1 191
AF227909.3 - 1 184
BC085725.1 - 1 284 7123652

View Full Product Details