Gene Transcripts

Gene Symbol
Smagp
Entrez Gene ID
300236
Gene Name
small cell adhesion glycoprotein
Gene Aliases
Chromosome Location
Chr. 7: 142244295 - 142260896 on Build Rnor_6.0
UniGene ID
Rn.162017
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_182817.1 NP_877969.1 4 301
XM_006242323.2 XP_006242385.1 4 430
GenBank mRNA AJ577837.1 - 4 301
BC101910.1 - 4 616 7933976

View Full Product Details