Gene Transcripts

Gene Symbol
Yipf5
Entrez Gene ID
361315
Gene Name
Yip1 domain family, member 5
Gene Aliases
Ac2-256
Chromosome Location
Chr. 18: 33401103 - 33436885 on Build Rnor_6.0
UniGene ID
Rn.7575
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001014150.3 NP_001014172.3 3 320
XM_006254664.2 XP_006254726.1 12 1481
GenBank mRNA AY321347.1 - 12 1487
BC083754.1 - 3 320

View Full Product Details