Gene Transcripts

Gene Symbol
Crlf3
Entrez Gene ID
54395
Gene Name
cytokine receptor-like factor 3
Gene Aliases
Cytor4
Chromosome Location
Chr. 10: 67383664 - 67401836 on Build Rnor_6.0
UniGene ID
Rn.203589
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001168612.1 NP_001162083.1 4 649
GenBank mRNA AF072835.1 - 557
CO555844.1 - 4 648
DY321186.1 - 4 642

View Full Product Details