Gene Transcripts

Gene Symbol
Il4r
Entrez Gene ID
25084
Gene Name
interleukin 4 receptor
Gene Aliases
Il4ra
Chromosome Location
Chr.1: 196942345 - 196967221 on Build Rnor_6.0
Uni Gene
Rn.10471
Interrogated Sequence Translated Protein
RefSeq NM_133380.2 NP_596871.2
XM_006230215.2 XP_006230277.1
XM_006230216.2 XP_006230278.1
XM_008759862.1 XP_008758084.1
XM_008759863.1 XP_008758085.1
XM_008759864.1 XP_008758086.1
GenBank mRNA AB015746.1 -
AB015747.1 -
AW527353.1 -
BQ204155.1 -
X69903.1 -

View Full Product Details