Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NR_026836.1 | 2 | 1012 | ||
NR_026837.1 | 1 | 1614 | |||
GenBank mRNA | AK095156.1 | - | 1 | 631 | |
BC040230.1 | - | 2 | 987 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AW902259.1 | - | 1 | 96 | Chr. 12 - 72255681 - 72271980 [-] |