Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000043.5 | NP_000034.1 | 9 | 3043 | |
NM_001320619.1 | NP_001307548.1 | 8 | 2960 | ||
NM_152871.3 | NP_690610.1 | 8 | 2980 | ||
NM_152872.3 | NP_690611.1 | 8 | 3018 | ||
NR_028033.3 | 7 | 2871 | |||
NR_028034.3 | 6 | 2733 | |||
NR_028035.3 | 7 | 2796 | |||
NR_028036.3 | 8 | 2934 | |||
NR_135313.1 | 7 | 2851 | |||
NR_135314.1 | 9 | 3034 | |||
NR_135315.1 | 7 | 2787 | |||
GenBank mRNA | AK026195.1 | - | 1 | 332 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BX093666.1 | - | 1 | 98 | Chr. 10 - 89016070 - 89016855 [+] |