Gene Transcripts
- Gene Symbol
- LOC253805
- Entrez Gene ID
- 253805
- Gene Name
- uncharacterized LOC253805
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Hs.667492
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC028622.2 |
- |
1 |
2487 |
|
BC050316.1 |
- |
1 |
2481 |
|
View Full Product Details