Gene Transcripts
- Gene Symbol
- HOXD11
- Entrez Gene ID
- 3237
- Gene Name
- homeobox D11
- Gene Aliases
- HOX4, HOX4F
- Chromosome Location
- Chr. 2: 176107356 - 176109588 on Build GRCh38
- UniGene ID
- Hs.421136
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK309681.1 |
- |
2 |
853 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AW630600.1 |
- |
1 |
93 |
|
Chr. 2 - 176109562 - 176109994 [+] |
View Full Product Details