Gene Transcripts
- Gene Symbol
- LOC101928907
- Entrez Gene ID
- 101928907
- Gene Name
- uncharacterized LOC101928907
- Gene Aliases
- Chromosome Location
- Chr. 15: 62060503 - 62062434 on Build GRCh38
- UniGene ID
- Hs.552949
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_135690.1 |
|
2 |
1286 |
|
GenBank mRNA |
BC034325.1 |
- |
1 |
1394 |
|
View Full Product Details