Gene Transcripts

Gene Symbol
TMEM185B
Entrez Gene ID
79134
Gene Name
transmembrane protein 185B
Gene Aliases
FAM11B
Chromosome Location
Chr. 2: 120217471 - 120223408 on Build GRCh38
UniGene ID
Hs.376722
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024121.2 NP_077026.2 1 4162
GenBank mRNA AK021716.1 - 1 896
AK090980.1 - 1 268

View Full Product Details