Gene Transcripts
- Gene Symbol
- Gm12603
- Entrez Gene ID
- 100040617
- Gene Name
- predicted gene 12603
- Gene Aliases
- OTTMUSG00000007693
- Chromosome Location
- Chr. 4: 89050307 - 89084610 on Build GRCm38
- UniGene ID
- Mm.171720
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_033533.1 |
|
3 |
579 |
|
GenBank mRNA |
AK144910.1 |
- |
3 |
579 |
|
BB819745.1 |
- |
3 |
404 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY734595.1 |
- |
3 |
577 |
|
Chr. 4 - 89050308 - 89084511 [+] |
GenBankmRNA |
CK623186.1 |
- |
3 |
471 |
|
Chr. 4 - 88956586 - 89084488 [+] |
View Full Product Details