Gene Transcripts
- Gene Symbol
- AU022793
- Entrez Gene ID
- 105976
- Gene Name
- expressed sequence AU022793
- Gene Aliases
- Chromosome Location
- Chr. 15: 39962649 - 39967515 on Build GRCm38
- UniGene ID
- Mm.394885
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_045719.1 |
|
3 |
1349 |
|
GenBank mRNA |
AK155489.1 |
- |
3 |
1352 |
|
View Full Product Details