Gene Transcripts
- Gene Symbol
- 5430425K12Rik
- Entrez Gene ID
- 71416
- Gene Name
- RIKEN cDNA 5430425K12 gene
- Gene Aliases
- Chromosome Location
- Chr. 13: 80940403 - 80948597 on Build GRCm38
- UniGene ID
- Mm.415500
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_103550.1 |
|
5 |
958 |
|
GenBank mRNA |
AK017339.1 |
- |
2 |
679 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY717153.1 |
- |
2 |
679 |
|
Chr. 13 - 80941059 - 80948576 [+] |
View Full Product Details