Gene Transcripts
- Gene Symbol
- A930036K24Rik
- Entrez Gene ID
- 77829
- Gene Name
- RIKEN cDNA A930036K24 gene
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Mm.134224
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK020935.1 |
- |
1 |
610 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AK044545.1 |
- |
|
637 |
|
Chr. - - [-] |
View Full Product Details