Gene Transcripts
- Gene Symbol
- D730047P14Rik
- Entrez Gene ID
- 77852
- Gene Name
- RIKEN cDNA D730047P14 gene
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK021347.1 |
- |
1 |
351 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY730940.1 |
- |
1 |
347 |
|
Chr. 9 - 78821880 - 78822371 [+] |
View Full Product Details