Gene Transcripts
- Gene Symbol
- Ints6
- Entrez Gene ID
- 18130
- Gene Name
- integrator complex subunit 6
- Gene Aliases
- 2900075H24Rik, AI480962, DICE1, Ddx26, HDB, Notch2l
- Chromosome Location
- Chr. 14: 62676325 - 62761370 on Build GRCm38
- UniGene ID
- Mm.319684
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK013790.1 |
- |
1 |
247 |
|
AK049304.1 |
- |
1 |
1857 |
|
AK169725.1 |
- |
1 |
1808 |
|
View Full Product Details