Gene Transcripts
- Gene Symbol
- 4930557J02Rik
- Entrez Gene ID
- 75357
- Gene Name
- RIKEN cDNA 4930557J02 gene
- Gene Aliases
- Chromosome Location
- Chr. 5: 34118740 - 34135339 on Build GRCm38
- UniGene ID
- Mm.410806
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_040703.1 |
|
2 |
731 |
|
GenBank mRNA |
AK016164.1 |
- |
2 |
733 |
|
View Full Product Details