Gene Transcripts
- Gene Symbol
- 4930447N08Rik
- Entrez Gene ID
- 71647
- Gene Name
- RIKEN cDNA 4930447N08 gene
- Gene Aliases
- Chromosome Location
- Chr. 3: 122795091 - 122801980 on Build GRCm38
- UniGene ID
- Mm.449992
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_045168.1 |
|
3 |
944 |
|
GenBank mRNA |
AK015409.1 |
- |
3 |
946 |
|
View Full Product Details