Gene Transcripts
- Gene Symbol
- LINC01099
- Entrez Gene ID
- 101928656
- Gene Name
- long intergenic non-protein coding RNA 1099
- Gene Aliases
- Chromosome Location
- Chr. 4: 177812046 - 177907936 on Build GRCh38
- UniGene ID
- Hs.508131
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_108093.1 |
|
2 |
628 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000514736.1 |
- |
2 |
628 |
|
Chr. 4 - 177903384 - 177907936 [-] |
View Full Product Details