Gene Transcripts
- Gene Symbol
- 4921507G05Rik
- Entrez Gene ID
- 66709
- Gene Name
- RIKEN cDNA 4921507G05 gene
- Gene Aliases
- Chromosome Location
- Chr. 12: 110840997 - 110843087 on Build GRCm38
- UniGene ID
- Mm.390504
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK014831.1 |
- |
|
137 |
|
AK015341.1 |
- |
1 |
103 |
|
View Full Product Details