Gene Transcripts

Gene Symbol
Tfb1m
Entrez Gene ID
224481
Gene Name
transcription factor B1, mitochondrial
Gene Aliases
AI429207
Chromosome Location
Chr. 17: 3519256 - 3557822 on Build GRCm38
UniGene ID
Mm.226554
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146074.1 NP_666186.1 1 58
GenBank mRNA AF508971.1 - 1 58
AK028212.1 - 1 79
BC032930.1 - 1 58 1382837

View Full Product Details