Gene Transcripts

Gene Symbol
Yipf1
Entrez Gene ID
230584
Gene Name
Yip1 domain family, member 1
Gene Aliases
C030002N13Rik
Chromosome Location
Chr. 4: 107314254 - 107359823 on Build GRCm38
UniGene ID
Mm.29118
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145550.3 NP_663525.1 4 387
GenBank mRNA AK081205.1 - 4 374
AK157536.1 - 4 374
BC009080.1 - 4 319 2655774

View Full Product Details