Gene Transcripts
- Gene Symbol
- Olr414
- Entrez Gene ID
- 56821
- Gene Name
- olfactory receptor 414
- Gene Aliases
- Gust43
- Chromosome Location
- Chr. 3: 20882324 - 20883259 on Build Rnor_6.0
- UniGene ID
- Rn.138116
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_020106.1 |
NP_064491.1
|
1 |
798 |
|
View Full Product Details