Gene Transcripts

Gene Symbol
Fgl2
Entrez Gene ID
84586
Gene Name
fibrinogen-like 2
Gene Aliases
Chromosome Location
Chr. 4: 10323598 - 10329241 on Build Rnor_6.0
UniGene ID
Rn.64635
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053455.2 NP_445907.2 2 940
GenBank mRNA AF323608.2 - 2 967
BC072502.1 - 2 889
BC085357.1 - 2 897
BC107665.1 - 2 913

View Full Product Details