Gene Transcripts

Gene Symbol
Klf4
Entrez Gene ID
114505
Gene Name
Kruppel-like factor 4 (gut)
Gene Aliases
GKLF
Chromosome Location
Chr. 5: 72283311 - 72287669 on Build Rnor_6.0
UniGene ID
Rn.7719
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053713.1 NP_446165.1 3 803
GenBank mRNA AF390546.1 - 3 803
BC085720.1 - 3 1196 7190219
FQ233252.1 - 1 170
FQ233720.1 - 1 170

View Full Product Details