Gene Transcripts

Gene Symbol
Klf4
Entrez Gene ID
114505
Gene Name
Kruppel-like factor 4 (gut)
Gene Aliases
GKLF
Chromosome Location
Chr. 5: 72283311 - 72287669 on Build Rnor_6.0
UniGene ID
Rn.7719
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053713.1 NP_446165.1 3 385
GenBank mRNA AF390546.1 - 3 385
BC085720.1 - 3 778 7190219

View Full Product Details