Gene Transcripts

Gene Symbol
Itgav
Entrez Gene ID
296456
Gene Name
integrin, alpha V
Gene Aliases
Cd51
Chromosome Location
Chr. 3: 71113267 - 71205958 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008762000.1 XP_008760222.1 15 2244

View Full Product Details