Gene Transcripts

Gene Symbol
F5
Entrez Gene ID
304929
Gene Name
coagulation factor V (proaccelerin, labile factor)
Gene Aliases
Ac2-120
Chromosome Location
Chr. 13: 82479997 - 82535540 on Build Rnor_6.0
UniGene ID
Rn.82910
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001047878.1 NP_001041343.1 12 3181
GenBank mRNA AY321333.1 - 12 3181

View Full Product Details