Gene Transcripts

Gene Symbol
Prc1
Entrez Gene ID
308761
Gene Name
protein regulator of cytokinesis 1
Gene Aliases
Chromosome Location
Chr. 1: 142087196 - 142108746 on Build Rnor_6.0
UniGene ID
Rn.138438
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107529.1 NP_001100999.1 12 1645
XM_006229433.2 XP_006229495.1 12 1657
XM_006229437.2 XP_006229499.1 12 1657

View Full Product Details