Gene Transcripts

Gene Symbol
H2-T9
Entrez Gene ID
15051
Gene Name
histocompatibility 2, T region locus 9
Gene Aliases
H-2T9
Chromosome Location
Chr. : - on Build
UniGene ID
Mm.195061
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010399.2 NP_034529.1 1024
GenBank mRNA BC044208.1 - 1035
M35248.1 - 1024

Gene Symbol
H2-T10
Entrez Gene ID
15024
Gene Name
histocompatibility 2, T region locus 10
Gene Aliases
H-2T10
Chromosome Location
Chr. 17: 36115871 - 36121444 on Build GRCm38
UniGene ID
Mm.426031
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010395.7 NP_034525.4 1038
GenBank mRNA AK088241.1 - 3, 4 540
AK153876.1 - 1027
AK161731.1 - 3, 4 1354
BC021751.1 - 1132 4165944
BC021760.1 - 1156 5055493
M35244.1 - 4, 5 1142
M35246.1 - 5, 6 1030

Gene Symbol
H2-T22
Entrez Gene ID
15039
Gene Name
histocompatibility 2, T region locus 22
Gene Aliases
27b, H-2T17, H-2T22, H2-T17, T22b
Chromosome Location
Chr. 17: 36037133 - 36042742 on Build GRCm38
UniGene ID
Mm.195061
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010397.4 NP_034527.1 4, 5 1143
GenBank mRNA AB359229.1 - 4, 5 888
AK133985.1 - 5, 6 1075
M35243.1 - 4, 5 1112
M35245.1 - 4, 5 1143
M35247.1 - 4, 5 1105

View Full Product Details