Gene Transcripts

Gene Symbol
CHP2
Entrez Gene ID
63928
Gene Name
calcineurin like EF-hand protein 2
Gene Aliases
Chromosome Location
Chr. 16: 23754627 - 23758951 on Build GRCh38
UniGene ID
Hs.178589
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022097.3 NP_071380.1 4, 5 773
GenBank mRNA AF146019.1 - 4, 5 778
AK290253.1 - 4, 5 359
AK313062.1 - 4, 5 629

View Full Product Details