Gene Transcripts

Gene Symbol
F8
Entrez Gene ID
2157
Gene Name
coagulation factor VIII
Gene Aliases
AHF, DXS1253E, F8B, F8C, FVIII, HEMA
Chromosome Location
Chr. X: 154835788 - 155022723 on Build GRCh38
UniGene ID
Hs.632836
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000132.3 NP_000123.1 23 6692
NM_019863.2 NP_063916.1 2 261
GenBank mRNA AK289947.1 - 2 191
AK295164.1 - 11 1489
AK313707.1 - 2 146
AX746360.1 - 23 6521
BC022513.1 - 2 180
BC064380.1 - 2 240
BC098389.1 - 2 259
BC111967.1 - 2 149
BC111969.1 - 2 148
K01740.1 - 23 6671
M14113.1 - 23 6692
M90707.1 - 2 156
X01179.1 - 23 6630

View Full Product Details