Gene Transcripts

Gene Symbol
Evx1
Entrez Gene ID
14028
Gene Name
even skipped homeotic gene 1 homolog
Gene Aliases
Evx-1
Chromosome Location
Chr. 6: 52313498 - 52318389 on Build GRCm38
UniGene ID
Mm.389754
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007966.4 NP_031992.1 1 424
GenBank mRNA BC126907.1 - 1 182
X54239.1 - 1 425

View Full Product Details