Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_010508.2 | NP_034638.2 | 11 | 1715 | |
GenBank mRNA | AK132431.1 | - | 11 | 1647 | |
AK136950.1 | - | 11 | 1691 | ||
AK141630.1 | - | 11 | 1651 | ||
AK148246.1 | - | 11 | 1657 | ||
AK157555.1 | - | 12 | 1743 | ||
AK178895.1 | - | 2 | 320 | ||
BC043935.1 | - | 11 | 1666 | 4507009 | |
BC052217.1 | - | 11 | 1675 | 30008497 | |
BC052429.1 | - | 11 | 1677 | 6836300 | |
M89641.1 | - | 11 | 1680 |