Gene Transcripts

Gene Symbol
HOXD11
Entrez Gene ID
3237
Gene Name
homeobox D11
Gene Aliases
HOX4, HOX4F
Chromosome Location
Chr. 2: 176107356 - 176109588 on Build GRCh38
UniGene ID
Hs.421136
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021192.2 NP_067015.2 2 836
GenBank mRNA AK309681.1 - 2 160
BC109394.1 - 3 338

View Full Product Details